Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hexanes, Isohexane
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:68…70 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–22 °C
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS06
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Boiling point:63 - 69 °C
- Evaporation residue:Max. 20 ppm
Specification Test Results
Frequently Bought Together