



n-Hexane ≥99.0% (by GC), Suprasolv® hypergrade for organic trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:–22 °C
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Purity (GC):≥99.0%
- Sum of Hexane Isomers+Methylcyclopentane (GC):≥99.9%
- Identity (IR):Conforms
- Evaporation Residue:≤3.0 mg/l
- Water:≤0.005%
- Color:≤10 Hazen
- GC/ECD (Retention Range 1,2,4-tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)):≤2 pg/ml
- GC/ECD (Retention range Dichloromethane to 1,2,4-Trichlorobenzene Individual Signals (Tetrachloromethane)):≤1 ng/ml
- GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)):≤2 ng/ml
- GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning Area 30 - 600 amu, Individual Signals (n- Tetradecane Standard):≤2 ng/ml
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Individual products and tailor-made chemicals for your technical applications Merck Pharma Segment Brochure Merck SupraSolv® High Purity Solvents for Gas Chromatography
Product Literature
msds_104369
SDS
ProductPage_104369
Product specification
Frequently Bought Together