Order Entry
Austria
ContactUsLinkComponent
n-Hexane ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
n-Hexane ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Certificates
undefined
n-Hexane ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Specification Test Results

  • Assay (GC):
    Min. 95 %
  • Non-volatile matter:
    max. 0.0005 %
  • Water (Karl Fischer):
    max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Autoignition Temperature:
    453 °F
  • Explosion Limit:
    7.7 %
  • Vapor Pressure:
    256 mmHg ( 37.7 °C)
  • Determination of P/N:
    ≤5 ng/L

Specification Test Results

Related Information

Frequently Bought Together