ContactUsLinkComponent
n-Hexane certified refence materials (ISO 17034)
  EHERC14195500
 :  
undefined
n-Hexane certified refence materials (ISO 17034)
  EHERC14195500
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070
  • Environmentally Preferable:
 : Hexyl hydride, Hexanen, Hexane
 :
 :86.18 g/mol
 :13,04712
 :69 °C (1013 hPa)
 :–94.3 °C
 :0.659 g/cm³ (20 °C)
 :–22 °C
 :Ambient
 :203-777-6
 :1208
 :3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :110-54-3
 :92112-69-1
 :H₃C(CH₂)₄CH₃
 :MFCD00009520