Xylene (mixture of isomers) ≥98.5% (by GC), AR® ACS for histology, Macron Fine Chemicals™
: Dimethylbenzene
:1330-20-7
:
:C₆H₄(CH₃)₂
:MFCD00077264
:106.17 g/mol
:13,10136
:140 °C (1013 hPa)
:–34 °C
:0.86…0.88 g/cm³ (20 °C)
:29 °C
:215-535-7
:1307
:3,III
:GHS02
:GHS07
:GHS08
:GHS09
:Ambient
- Environmentally Preferable:
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- ACS - Assay (m,p,o-xylenes and ethylbenzene) (by GC):≥98.5 %
- ACS - Color (APHA):≤10
- ACS - Ethylbenzene (C₆H₅C₂H₅):≤25 %
- ACS - Particulate Contamination:Passes Test
- ACS - Residue after Evaporation:≤0.002 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Sulfur Compounds (as S):≤0.0005 %
- ACS - Water (H₂O):≤0.02 %
- Appearance (clear, colorless liquid):—
Frequently Bought Together