



n-Hexane, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8205.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (n-hexane):min. 99 %
- Assay (sum of 5 isomers, total hexanes plus methylcyclopentane):min. 99 %
- Aromatic Compounds (as C6H6):max. 0.02 %
- Color (APHA):max. 10
- Residue after Evaporation:max. 5 ppm
- Sulfur Compounds (as S):max. 0.005 %
- Thiophene:passes test
- Water Soluble Titrable Acid (meq/g):max. 0.0002
- at 210 nm:max. 1.00
- at 220 nm:max. 0.20
- at 230 nm:max. 0.10
- at 240 nm:max. 0.04
- at 250 nm:max. 0.02
- at 280-400 nm:max. 0.01
Specification Test Results
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520