Hexane (mixture of isomers) 99% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hexanes, Isohexane
CAS Number:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.659 g/cm³
Flash Pt:–22 °C
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- GC:≥98.5 % Sum of 5 isomers + total Hexanes + Methylcyclopentane
- UV:at 400 nm A: ≤0.01
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Sulfur compounds (as S):≤0.005 %
- Water soluble titr. acid:≤0.0003 meq/g
- Thiophene:passes test
Specification Test Results
Frequently Bought Together