Order Entry
Austria
ContactUsLinkComponent
n-Hexane 97% for electronic use (MOS) residue-free
n-Hexane 97% for electronic use (MOS) residue-free
Catalog # ACRO327890010
CAS Number:  
undefined
n-Hexane 97% for electronic use (MOS) residue-free
Catalog # ACRO327890010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.01 % (Coulometric)
  • GC:
    ≥96 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity:
    ≤0.002 %
  • Chloride (Cl):
    ≤2 ppm
  • Arsenic (As):
    ≤10 ppb
  • Barium (Ba):
    ≤20 ppb
  • Copper (Cu):
    ≤10 ppb
  • Iron (Fe):
    ≤50 ppb
  • Heavy metals (as Pb):
    ≤1 ppm

Specification Test Results

About this item

Synonyms: Hexyl hydride, Hexanen, Hexane
 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.659 g/cm³
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS07
 :GHS08
 :GHS09
 :GHS02
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520