n-Hexane 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
: Hexyl hydride, Hexanen, Hexane
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD00009520
:86.18 g/mol
:13,04712
:69 °C (760 mmHg)
:–95 °C
:0.659 g/cm³
:–22 °C
:203-777-6
:1208
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Environmentally Preferable:
- Appearance:Clear liquid
- Water:≤0.005 % (Coulometric)
- GC:≥95 %
- UV:at 200 nm A: ≤0.602
- Color scale:≤5 APHA
- Residue after evaporation:≤0.0002 %
- Acidity (CH3COOH):≤0.001 %
Frequently Bought Together