ContactUsLinkComponent
n-Hexane ∼95% (n-hexane) for pesticide residue analysis
n-Hexane ∼95% (n-hexane) for pesticide residue analysis
Certificates
undefined
n-Hexane ∼95% (n-hexane) for pesticide residue analysis
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.01 %
  • GC:
    (ECD) Passes test, (NPD) Passes test, (FID) Passes test
  • Residue after evaporation:
    ≤2 ppm