Hexane (mixture of isomers), technical
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hexanes, Isohexane
CAS Number:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Boiling Pt:69 °C
Melting Pt:–95 °C
Density:0.659 g/cm³
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:295-570-2
UN:1208
ADR:3,II
REACH:01-2119484651-34
:GHS02
:GHS07
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
Specification Test Results
Frequently Bought Together