Order Entry
Austria
ContactUsLinkComponent
n-Hexadecane, TECHNICAL
n-Hexadecane, TECHNICAL
Catalog # 24556.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
n-Hexadecane, TECHNICAL
Catalog # 24556.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

Synonyms: Hexadecane, Cetane
 :
 :226.45 g/mol
Boiling Pt:287 °C (1013 hPa)
Melting Pt:18 °C
Density:0.77331 g/cm³ (20 °C)
Flash Pt:135 °C (closed cup)
Storage Temperature:Ambient
EINECS:208-878-9
 :GHS08
CAS Number:544-76-3
Formula:H₃C(CH₂)₁₄CH₃
MDL Number:MFCD00008998