



n-Heptane ≥99%, GPR RECTAPUR®
:142-82-5
:
:H₃C(CH₂)₅CH₃
:MFCD00009544
:100.20 g/mol
:13,04679
:98.4 °C (1013 hPa)
:–90,6 °C
:0.6838 g/cm³ (20 °C)
:–4 °C
:205-563-8
:1206
:3,II
:01-2119457603-38
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
- Assay:Min. 99 %
- IR Spectrum:Passes test
- Boiling point:98 - 99 °C
- Density (20/4):0.681 - 0.685
- Acidity:Max. 0.0008 meq/g
- Substances coloured by H2SO4:Max. 90 APHA
- Evaporation residue:Max. 20 ppm
1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Tin bottle for liquids UN Y (ZPACK12321007)
Packaging Information
spec_VWRC24549.292 spec_VWRC24549.326 spec_VWRC24549.326 spec_VWRC24549.361 spec_VWRC24549.361
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together