



n-Heptane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
:142-82-5
:
:H₃C(CH₂)₅CH₃
:MFCD00009544
:100.20 g/mol
:13,04679
:98.4 °C (1013 hPa)
:–90.6 °C
:0.6838 g/cm³ (20 °C)
:–4 °C
:205-563-8
:1206
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
- Assay (by GC):min. 99.0 %
- Boiling Range:97.5 - 99.0 °C
- Color (APHA):max. 10
- Recorded Boiling Point:98.4 °C
- Residue after Evaporation:max. 0.001 %
- Total Sulfur Compounds (as S):max. 0.005 %
- Water (H2O):max. 0.01 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Frequently Bought Together