Order Entry
Austria
ContactUsLinkComponent
n-Heptane 99+% for residue analysis, for trace analysis of polyaromatic hydrocarbons
Irritant
Health
Environment
Flammable
n-Heptane 99+% for residue analysis, for trace analysis of polyaromatic hydrocarbons
Certificates
undefined
Irritant
Health
Environment
Flammable
n-Heptane 99+% for residue analysis, for trace analysis of polyaromatic hydrocarbons

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.001 % (Coulometric)
  • GC:
    ≥99 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0003 %
  • Acidity:
    ≤0.002 %

Specification Test Results

Related Information

Frequently Bought Together