Order Entry
Austria
ContactUsLinkComponent
n-Dodecane ≥99% for synthesis, Sigma-Aldrich®
Health
n-Dodecane ≥99% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Health
n-Dodecane ≥99% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (GC, area%):
    ≥ 99.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    0.748 - 0.749
  • Identity (IR):
    passes test

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together