Order Entry
Austria
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS), BAKER, Laboratory Reagent, J.T.Baker®
Flammable
Toxic
Sodium dodecyl sulphate (SDS), BAKER, Laboratory Reagent, J.T.Baker®
Artikelnr. 2811.1000
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Toxic
Sodium dodecyl sulphate (SDS), BAKER, Laboratory Reagent, J.T.Baker®
Artikelnr. 2811.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    min. 95 %

Specification Test Results

About this item

 :
 :288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
ADR:4.1,II
 :GHS02
 :GHS06
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175