Order Entry
Austria
ContactUsLinkComponent
 
Sodium dodecyl sulphate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free
 
undefined
Sodium dodecyl sulphate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥99.0 %
  • UV:
    (0.1 M in water)
  • Dnase,Rnase,Protease activity:
    None detected
  • Chloride (Cl):
    ≤1000 ppm
  • Phosphate (PO4):
    ≤5 ppm
  • Copper (Cu):
    ≤5 ppm
  • Lead (Pb):
    ≤5 ppm

 

Frequently Bought Together