Order Entry
Austria
ContactUsLinkComponent
 
Glycerine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
    
 
undefined
Glycerine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)

 

 

  • Assay:
    min. 99.5 %
  • Silver Reducing Substances:
    passes test
  • Color:
    passes test
  • Density (g/ml) at 25°C:
    min. 1.255
  • Fatty Acid Esters:
    max. 0.05 %
  • Neutrality:
    passes test
  • Residue after Ignition:
    max. 0.005 %
  • Substances Darkened by H2SO4:
    passes test
  • Sulfate (SO4):
    max. 0.001 %
  • Chloride (Cl):
    max. 5
  • Heavy Metals (as Pb):
    max. 2

 

 

Frequently Bought Together