

Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:107-21-1
:
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:203-473-3
REACH:01-2119456816-28
:GHS07
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.7 %
- Appearance of soln. (50 % V/V; water):Passes test
- IR Spectrum:Passes test
- Acidity:Max. 0.0001 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Colouration:Max. 10 APHA
- Density (20/4):1.111 - 1.115
- Substances coloured by H2SO4:Max. 150 APHA
- Formaldehyde:Max. 20 ppm
- Heavy metals (as Pb):Max. 1 ppm
- Ignition residue (SO4):Max. 30 ppm
- Substances reducing KMnO4 (as O):Max. 3 ppm
- Water:Max. 0.1 %
- Cl (Chloride):Max. 0.2 ppm
- SO4 (Sulphate):Max. 20 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 0.5 ppm
- Pb (Lead):Max. 1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
SDS_EN_CH_VWRC24041.pdf SDS_EN_DE_VWRC24041.pdf SDS_EN_NL_VWRC24041.pdf SDS_EN_UK_VWRC24041.pdf sds_VWRC24041
SDS
Frequently Bought Together