Ethylene glycol, EMPLURA®, Supelco®
: Merck
:107-21-1
:
:HOCH₂CH₂OH
:MFCD00002885
:62.07 g/mol
:13,03832
:196…198 °C (1013 hPa)
:–13 °C
:1.115 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:116 °C
:Ambient
:203-473-3
:01-2119456816-28
:GHS08