Order Entry
Austria
ContactUsLinkComponent
Ethylene glycol, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Ethylene glycol, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Ethylene glycol, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.0 %
  • Acidity (as CH3COOH):
    max. 0.01 %
  • Boiling Range:
    194.0 - 200.0 °C
  • Boiling Point:
    195 - 198 °C
  • Color (APHA):
    max. 10
  • Density (g/ml) at 20°C:
    1.110 - 1.114
  • Residue after Ignition:
    max. 0.005 %
  • Water (H2O):
    max. 0.2 %
  • Chloride (Cl):
    max. 5
  • Iron (Fe):
    max. 0.2

Specification Test Results

Related Information

Frequently Bought Together