Ethylene glycol, high purity
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:107-21-1
:
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:196 – 198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:203-473-3
REACH:01-2119456816-28
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Acidity:≤0.002%
- Iron:≤0.0005%
- Moisture (KF):≤0.5%
- Purity:≥99.0 %
Specification Test Results
Frequently Bought Together