

Ethylene glycol 99+%, extra pure
:107-21-1
:
:HOCH₂CH₂OH
:MFCD00002885
:62.07 g/mol
:13,03832
:196…198 °C (1013 hPa)
:–13 °C
:1.113 g/cm³
:116 °C
:Ambient
:203-473-3
:01-2119456816-28
:GHS07
:GHS08
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4300 to 1.4330 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- Sulfated ash:≤0.05 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Acidity (CH3COOH):≤0.002 %
Frequently Bought Together