Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:11 °C (closed cup)
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (GC):Min. 99.8 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0003 meq/g
- Evaporation residue:Max. 5 ppm
- Water:Max. 0.05 %
- Absorbance (220 nm):Max. 0.4
- Absorbance (240 nm):Max. 0.15
- Absorbance (260 nm):Max. 0.02
- Transmittance (220 nm):Min. 45 %
- Transmittance (240 nm):Min. 75 %
- Transmittance (260 nm):Min. 96 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC20837.320 spec_VWRC20837.360 spec_VWRC20837.9030
Product specification
Frequently Bought Together