


Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
:M-Clarity™ quality level = MQ100
Flash Pt:11 °C (closed cup)
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-98 °C
- Molar Mass:32.04 g/mol
- Boiling Point:64.5 °C (1013 hPa)
- Vapor Pressure:128 hPa (20 °C)
- Flash Point:10 °C
- Refractive Index:1.33
- Explosion Limit:5.5 - 44 %(V)
- Density:0.792 g/cm3 (20 °C)
- Ignition Point:455 °C DIN 51794
- Synonyms:MeOH, Hydroxymethane, Methyl alcohol, Carbinol
- Hill Formula:CH4O
- Chemical Formula:CH3OH
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06035.1000_TDS_EN MERC1.06035.2500_TDS_EN ProductPage_106035
Product specification
Merck Pharma Segment Brochure
Product Literature
msds_106035
SDS
Frequently Bought Together