

Methanol ≥99.9% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS08
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Suitability for residue analysis:Max. 5 ng/l
- Vapor Pressure:97.68 mmHg ( 20 °C)
- Refractive Index:n20/D 1.329 (lit.)
- Explosion Limit:36 %
- Autoignition Temperature:725 °F
Specification Test Results
Frequently Bought Together