Order Entry
Austria
ContactUsLinkComponent
 
Methanol ≥99.8%, Ultrapure for HPLC
Catalog # 22909.K2
   Safer Human and Environmental Health
CAS Number:  
undefined
Methanol ≥99.8%, Ultrapure for HPLC
Catalog # 22909.K2
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

About this item

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
CAS Number:67-56-1
 :
Formula:CH₃OH
MDL Number:MFCD00004595
 :32.04 g/mol
Merck Index:14,5957
Boiling Pt:64.7 °C
Melting Pt:–98 °C
Density:0.791
Flash Pt:11 °C (52 °F)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08