

Ethanol absolute (untaxed) ≥99.8% (by GC), CHROMASOLV™ A15 for HPLC, without additive, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Merck Index:12,03806
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.790…0.791 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Vapor Density:1.59 (vs air)
- Assay (GC):Min. 99.8 %
- Refractive index (n 20/D):1.360 - 1.362
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.2 %
- Absorbance at 260 nm:Max. 0.01
- Absorbance at 240 nm:Max. 0.1
- Absorbance at 205 nm:Max. 1.0
- Suitability test (HPLC):complying
- Autoignition Temperature:683 °F
- Refractive Index:n20/D 1.361
- Explosion Limit:19 %, 60 °F
Specification Test Results
Frequently Bought Together