Order Entry
Austria
ContactUsLinkComponent
Ethanol absolute ≥99.5%, TechniSolv®, pure
Flammable
Irritant
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Assay (V/V):
    Min. 99.5 %
  • Density (20/20):
    0.788 - 0.794
  • Water:
    Max. 0.5 %

Specification Test Results

Frequently Bought Together