Order Entry
Austria
ContactUsLinkComponent
Ethanol absolute ≥99.5%, GPR RECTAPUR®
Flammable
Irritant
Ethanol absolute ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
Ethanol absolute ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99,5 %
  • Assay (V/V):
    Min. 99,8 %
  • IR Spectrum:
    Passes test
  • Refractive index (20°C):
    1,360 - 1,.362
  • Evaporation residue:
    Max. 20 ppm
  • Water (KF):
    Max. 0,2%
  • Free acid (as CH₃COOH):
    Max. 0,001%
  • Aldehydes (as CH₃CHO):
    Max. 0,005%
  • Free alkali (as NH₃):
    Max. 0,0005%
  • Methanol (GC):
    Max. 0,1%
  • S-compounds (as S):
    Max. 5 ppm

Specification Test Results

Frequently Bought Together