Orders LinkContactUsLinkComponent
Dispersers, LABOR-PILOT 2000/4
Supplier:   IKA
Dispersers, LABOR-PILOT 2000/4
Supplier:  IKA
Certificates
Dispersers, LABOR-PILOT 2000/4
Dispersers, LABOR-PILOT 2000/4
5 Options Available Below
Product Details & Documents
The LABOR-PILOT is a compact and multi-purpose inline laboratory dispersing machine. It is designed for solid-liquid mixing, homogenising, emulsifying, suspending and wet milling. The LABOR-PILOT 2000/4 is suitable for wide range of applications, particularly in the chemical, cosmetic, pharmaceutical, plastics, colours and food industries. It is uniquely designed with advanced technology and is available with six interchangeable modules.
  • Flexibility and ease of use: One machine suits for many applications and variety of processes
  • Together with inline machines of 2000 series enables seamless transition of processes from laboratory to production while retaining the product quality
  • Easy and quick exchange of the modules as well as conversion to a complete working plant
  • CIP and SIP capable
  • Integral electronic control via LABOR-PILOT-CONTROLLER
It can be customized to fit particular procedural demands. Important process parameters such as circumferential speed, tools and mixing chamber geometry are structurally identical to those of the IKA 2000 inline production machines: UTL, DR, DRS, MHD, MK and MKO. Therefore, the LABOR-PILOT ensures a reliable scale-up by offering the possibility to work with the same method from formulation development to mass production. The laboratory dispersing machine LABOR-PILOT 2000/4 can be upgraded to a high-speed dispersing machine with the LABOR-PILOT-CONTROLLER.
The LABOR-PILOT 2000/4 differs from the alternative PROCESS-PILOT 2000/4 machine due to the applied seal technology and two additional process modules for powder entry in recirculation mode. For working under pressure/vacuum and with elevated temperatures or abrasive materials, we recommend the machine PROCESS-PILOT. The basic inline machine is equipped with the single-stage dispersing module ULTRA-TURRAX® UTL. It is best suited for batch applications and can be adapted for the manufacturing of dispersions from coarse to fine. Compared to a conventional stirrer, the mixing time is reduced by up to 50%. Depending on the application, the basic unit can be modified quickly with different working modules.
The DISPAX-REACTOR® DR module converts the LABOR-PILOT into a multi-stage high capacity dispersing machine for fine particle size reduction. With just one pass, the module DR does typically achieve fine dispersions with an extremely narrow particle size distribution. It also guarantees an optimum homogeneity, which is an important pre-condition for the long-time stability of emulsions or suspensions.
The MHD module is designed for a fully continuous incorporation of powders into liquids. By use of this module, the mixing relation of solid and liquid should be volumetrically or gravimetrically controlled. Depending on the product characteristics, dispersions with viscosities up to 50 Pas or solid contents up to 80% can be achieved just after one pass.
The Colloid Mill MK and Cone Mill MKO modules are used for wet or fine milling of hard and granular raw materials or for the production of finest emulsions as well as for improving of quality of finished dispersions. The MK module consists of two cones with differently directed flow channels on their surfaces. The cones can be shifted against each other to a minimum distance, thus varying friction as well as flow capacity. By adjusting the milling gap, the achievable particle size can be controlled. The MKO module works on the same principle as the MK module except the outlet area of its cones is coated with abrasion-resistant tungsten carbide. This results in an enlarged friction surface that enables to produce even finer suspensions at reduced flow.
Product Family Options
LABOR-PILOT Controller
Product Information
  • Availability
    Price
  • IKAAT054916
    Controller, 1,5 kW, LABOR-PILOT
    Controller, 1,5 kW, LABOR-PILOT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Modules
Product Information
  • Availability
    Price
  • IKAAT054917
    MK module for LABOR-PILOT
    MK module for LABOR-PILOT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • IKAAT055013
    DISPAX-REACTOR® module for LABOR-PILOT
    DISPAX-REACTOR® module for LABOR-PILOT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • IKAAT055142
    MHD module for LABOR-PILOT
    MHD module for LABOR-PILOT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • IKAAT061069
    MKO module for LABOR-PILOT
    MKO module for LABOR-PILOT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings standard for module UTL
    Sealing set O-rings standard for module UTL
    IKAAT056678
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Hose nozzle DN 15 for LABOR-PILOT
    Hose nozzle DN 15 for LABOR-PILOT
    IKAAT056486
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Three way stop cock for LABOR-PILOT
    Three way stop cock for LABOR-PILOT
    IKAAT056482
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Blind nut DN 15 for LABOR-PILOT
    Blind nut DN 15 for LABOR-PILOT
    IKAAT056484
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Two way ball valve for LABOR-PILOT
    Two way ball valve for LABOR-PILOT
    IKAAT056468
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Telescopic stand for LABOR-PILOT
    Telescopic stand for LABOR-PILOT
    IKAAT055330
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings of FFPM / FEP for module DISPAX-REACTOR®
    Sealing set O-rings of FFPM / FEP for module DISPAX-REACTOR®
    IKAAT056688
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Generator, 2P, for module UTL
    Generator, 2P, for module UTL
    IKAAT055002
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Generator, 2G, for module UTL
    Generator, 2G, for module UTL
    IKAAT055001
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Generator, 6F, for module UTL
    Generator, 6F, for module UTL
    IKAAT055000
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Generator, 4M for module UTL
    Generator, 4M for module UTL
    IKAAT054949
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Funnel for LABOR-PILOT
    Funnel for LABOR-PILOT
    IKAAT056458
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set for circulation bend for LABOR-PILOT
    Sealing set for circulation bend for LABOR-PILOT
    IKAAT056730
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings of FFPM / FEP for module MKO
    Sealing set O-rings of FFPM / FEP for module MKO
    IKAAT061084
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Circulating bend with intermediate piece for LABOR-PILOT
    Circulating bend with intermediate piece for LABOR-PILOT
    IKAAT057796
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Anchor stirrer for LABOR-PILOT
    Anchor stirrer for LABOR-PILOT
    IKAAT058564
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Wipers for anchor stirrer, PTFE, for LABOR-PILOT
    Wipers for anchor stirrer, PTFE, for LABOR-PILOT
    IKAAT058714
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Standard sealing set O-rings, Viton®, for module MKO
    Standard sealing set O-rings, Viton®, for module MKO
    IKAAT061081
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings FFPM / FEP for module UTL
    Sealing set O-rings FFPM / FEP for module UTL
    IKAAT056680
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings standard of Viton® for module DISPAX-REACTOR®
    Sealing set O-rings standard of Viton® for module DISPAX-REACTOR®
    IKAAT056687
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings of FFPM / FEP for module MK
    Sealing set O-rings of FFPM / FEP for module MK
    IKAAT056694
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Spiral gearing milling tool for module MK
    Spiral gearing milling tool for module MK
    IKAAT056698
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings of FFPM / FEP for module MHD
    Sealing set O-rings of FFPM / FEP for module MHD
    IKAAT056691
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings standard of Viton® for module MK
    Sealing set O-rings standard of Viton® for module MK
    IKAAT056693
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Sealing set O-rings standard of Viton® for module MHD
    Sealing set O-rings standard of Viton® for module MHD
    IKAAT056689
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Shaft sealing rig, PTFE, for modules DISPAX-REACTOR®, UTL,MHD, MK and MKO
    Shaft sealing rig, PTFE, for modules DISPAX-REACTOR®, UTL,MHD, MK and MKO
    IKAAT054617
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Milling tool WCCO-coated for module MKO
    Milling tool WCCO-coated for module MKO
    IKAAT061389
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Hose nozzle K / M DN 25 for continuous operation of LABOR-PILOT
    Hose nozzle K / M DN 25 for continuous operation of LABOR-PILOT
    IKAAT057251
    Log in for availability
    1 * 1 ST