Orders LinkContactUsLinkComponent
Surface protection papers, LabSorb
Surface protection papers, LabSorb
   Sustainable Options Available
Supplier:   Sartorius
Surface protection papers, LabSorb
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Surface protection papers, LabSorb
7 Options Available Below
Product Details & Documents
The LabSorb protection papers are highly absorptive papers coated on one side with polyethylene.
  • Prevent radioactive contamination of work surfaces (in radiochemical laboratories)
  • Protect laboratory bench surfaces from spillage or splashes of liquids by preventing absorption and seepage of these liquids
  • Line animal cages for protection and improvement of hygiene
  • Protect objects from falling on hard surfaces: reduce impact
Used with the cellulose side up, the paper absorbs liquids, which are stopped by their polyethylene layer and thus prevented from soaking through. Used with the polyethylene side up, the papers are highly useful for recovering valuable or toxic liquids.
Basis weight: 140 g/m²; Absorption: 150%
Specifications for Product Family
Specifications
  • Catalog No:
  • SARTFT2601-460570K
  • 515-1633
  • SARTFT1-601-460050
  • 515-1629
  • 515-1628
  • 515-1630
  • SARTFT1-601-600100
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifications
Product Family Options
Product Information
  • Colour
    Packed
    L×W
    Availability
    Price
  • White
    Sheets
    460×570 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 174,00 €
    174,00 €
    1 * 50 ST
       174,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT2601-460570K
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • White
    Sheets
    480×600 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 184,00 €
    184,00 €
    1 * 50 ST
       184,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1633
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • White
    Roll
    460×50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 214,00 €
    214,00 €
    1 * 1 ST
       214,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT1-601-460050
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • White
    Roll
    400×50 000 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 230,00 €
    230,00 €
    1 * 1 Rolle
       230,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1629
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • White
    Roll
    600×50 000 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 323,00 €
    323,00 €
    1 * 1 Rolle
       323,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1628
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • White
    Roll
    400×100 000 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 411,00 €
    411,00 €
    1 * 1 Rolle
       411,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1630
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • White
    Roll
    600×100 000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 576,00 €
    576,00 €
    1 * 1 ST
       576,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT1-601-600100
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact