Order Entry
Austria
ContactUsLinkComponent
Ethanol absolute Ph. Eur., J.T.Baker®
Flammable
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
Flammable
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Assay (C2H5OH):
    min. 99.5 % (v/v)
  • Acidity or Alkalinity (as CH3COOH):
    max. 30 ppm
  • Appearance:
    passes test
  • Identification:
    passes test
  • Relative Density:
    0.790 - 0.793
  • Residue on Evaporation:
    max. 25 ppm
  • at 240 nm:
    max. 0.40
  • at 250-260 nm:
    max. 0.30
  • at 270-340 nm:
    max. 0.10
  • Curve is smooth:
    passes test
  • Acetaldehyde + Acetal:
    max. 10 ppm (v/v)
  • Benzene:
    max. 2 ppm (v/v)
  • Methanol (CH3OH):
    max. 200 ppm (v/v)
  • Total of other Impurities (based on surface area of internal standard 4-Methylpentan-2-ol):
    max. 300 ppm

Specification Test Results

Related Information

Frequently Bought Together