2-Propanol
Marketsource
Supplier: Roth Carl
About this item
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0.786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
6 Options Available Below
Product Details & Documents
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0.786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ROTH6752.1
- ROTH6752.3
- ROTH6752.2
- ROTH6752.4
- ROTH6752.5
- ROTH6752.6
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default
- Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default
- Chem21 Solvent Selection Guide recommended by default
- Chem21 Solvent Selection Guide recommended by default
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH6752.1Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health Marketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH6752.3Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by defaultSafer Human and Environmental Health Marketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH6752.2Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health Marketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH6752.4Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by defaultSafer Human and Environmental Health Marketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH6752.5Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by defaultSafer Human and Environmental Health Marketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH6752.6Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by defaultSafer Human and Environmental Health Marketsource



