

2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
EINECS:200-661-7
UN:1219
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:12 °C
REACH:01-2119457558-25
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.8 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Evaporation residue:Max. 5 ppm
- Water:Max. 0.1 %
- Absorbance (220 nm):Max. 0.3
- Absorbance (230 nm):Max. 0.1
- Absorbance (250 nm):Max. 0.02
- Transmittance (220 nm):Min. 50 %
- Transmittance (230 nm):Min. 80 %
- Transmittance (250 nm):Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 4L Glass bottle DIN 38 (ZPACK13415438)
Packaging Information
20880_Propan-2-Ol_BSE_rev0
BSE/TSE Certificates
spec_VWRC20880.290 spec_VWRC20880.320 spec_VWRC20880.400 spec_VWRC20880.400 spec_VWRC20880.400
Product specification
More
Frequently Bought Together