Order Entry
Austria
ContactUsLinkComponent
 
2-Propanol ≥99.5% (by GC), EMPARTA® ACS analytical reagent, Supelco®
 :  Merck
    
 
undefined
2-Propanol ≥99.5% (by GC), EMPARTA® ACS analytical reagent, Supelco®
 :  Merck
    
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -89.5 °C
  • Molar Mass:
    60.1 g/mol
  • Boiling Point:
    82.4 °C (1013 hPa)
  • Vapor Pressure:
    43 hPa (20 °C)
  • Flash Point:
    12 °C
  • Refractive Index:
    1.378
  • Explosion Limit:
    2 - 13.4 %(V)
  • Density:
    0.786 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral
  • Ignition Point:
    425 °C
  • Evaporation Number:
    11

 

Frequently Bought Together