

2-Propanol, LiChrosolv®, gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
EINECS:200-661-7
UN:1219
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:12 °C
REACH:01-2119457558-25
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Related Information
Certificates
Search by Lot NumberMERC1.01040.2500_TDS_EN ProductPage_101040 ProductPage_101040
Product specification
Merck Pharma Segment Brochure
Product Literature
msds_101040
SDS
Frequently Bought Together