2-Propanol, SupraSolv® for gas chromatography, Supelco®
: Merck
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:200-661-7
:1219
:3,II
:M-Clarity™ quality level = MQ100
:12 °C
:01-2119457558-25
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Solubility:(20 °C) soluble
- Melting Point:-89.5 °C
- Molar Mass:60.1 g/mol
- Boiling Point:82.4 °C (1013 hPa)
- Vapor Pressure:43 hPa (20 °C)
- Flash Point:12 °C
- Refractive Index:1.378
- Explosion Limit:2 - 13.4 %(V)
- Density:0.786 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:425 °C
- Evaporation number:11
- Water absorption:1000 g/kg
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together