2-Propanol, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
EINECS:200-661-7
UN:1219
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:12 °C
REACH:01-2119457558-25
:GHS02
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-89.5 °C
- Molar Mass:60.1 g/mol
- Boiling Point:82.4 °C (1013 hPa)
- Vapor Pressure:43 hPa (20 °C)
- Flash Point:12 °C
- Refractive Index:1.378
- Explosion Limit:2 - 13.4%(V)
- Density:0.786 g/cm³ (20 °C)
- pH:(H₂O, 20 °C) neutral
- Ignition Point:425 °C
- Evaporation Number:11
- Water Absorption:1000 g/kg
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_100993
SDS
ProductPage_100993
Product specification
Solvents for spectroscopy Uvasol
Product Literature
Frequently Bought Together