Order Entry
Austria
ContactUsLinkComponent
2-Propanol standard for the analysis of tobacco, Supelco®
Flammable
Irritant
2-Propanol standard for the analysis of tobacco, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
2-Propanol standard for the analysis of tobacco, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -89 °C
  • Boiling Point:
    82 °C
  • Vapor Pressure:
    41.6 hPa
  • Flash Point:
    12 °C
  • Refractive Index:
    1.378
  • Explosion Limit:
    2 - 12 %(V)
  • Density:
    0.78 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    425 °C
  • Evaporation number:
    2.93
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information

Frequently Bought Together