Order Entry
Austria
ContactUsLinkComponent
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Flammable
Irritant
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99 %
  • Acidity (as CH3COOH):
    max. 0.002 %
  • Alkalinity (as NH3):
    max. 0.002 %
  • Boiling Range:
    max. 1.0 °C
  • Density (g/ml) at 25°C:
    0.781 - 0.783
  • Recorded Boiling Point:
    82 °C
  • Residue after Evaporation:
    max. 0.005 %
  • Water (H2O):
    max. 0.2 %

Specification Test Results

Related Information

Frequently Bought Together