Order Entry
Austria
ContactUsLinkComponent
1-Tetradecanol for synthesis, Sigma-Aldrich®
1-Tetradecanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1-Tetradecanol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    36 - 39 °C
  • Molar Mass:
    214.39 g/mol
  • Boiling Point:
    277 - 288 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    145 °C
  • Refractive Index:
    1.4346 (50 °C, 589 nm)
  • Density:
    0.8 g/cm3 (60 °C)
  • Ignition Point:
    240 °C

Specification Test Results

Related Information

Frequently Bought Together