


1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:71-23-8
:
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941
:60.1 g/mol
Merck Index:13,07934
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–127 °C
Density:0.8053 g/cm³ (20 °C. DIN 51757)
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:200-746-9
UN:1274
ADR:3,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Assay (on anhydrous substance):Min. 99.5 %
- Distillation range:Passes test Reag.Ph.Eur.
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0006 meq/g
- Boiling point:96.9 - 97.5 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.801 - 0.804
- Density (20/20):0.802 - 0.806
- Acetone:Max. 100 ppm
- Evaporation residue:Max. 10 ppm
- Ethanol:Max. 0.05 %
- Methanol:Max. 0.05 %
- Propan-2-ol:Max. 0.05 %
- Propionaldehyde:Max. 0.06 %
- Water:Max. 0.1 %
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.05 ppm
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC20861.294 spec_VWRC20861.294 spec_VWRC20861.320 spec_VWRC20861.320 spec_VWRC20861.363
Product specification
More
Frequently Bought Together