1-Propanol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:71-23-8
:
:H₃CCH₂CH₂OH
:MFCD00002941
:60.1 g/mol
:13,07934
:97 °C (1013 hPa)
:–127 °C
:0.8053 g/cm³ (20 °C)
:200-746-9
:1274
:3,II
:M-Clarity™ quality level = MQ300
:15 °C
:GHS02
:GHS05
:GHS07
:Ambient
- Solubility:(20 °C) miscible in all proportions
- Melting Point:-127 °C
- Molar Mass:60.1 g/mol
- Boiling Point:96.5 - 98 °C (1013 hPa)
- Vapor Pressure:19 hPa (20 °C)
- Flash Point:15 °C
- Explosion Limit:2.1 - 19.2 %(V)
- Density:0.80 g/cm3 (20 °C)
- pH:7 (200 g/l, H2O, 20 °C)
- Ignition Point:360 °C
- Evaporation number:17
Frequently Bought Together