1-Propanol 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-23-8
:
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941
:60.1 g/mol
Merck Index:13,07934
Boiling Pt:97 °C (760 mmHg)
Melting Pt:–127 °C
Density:0.8 g/cm³
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:200-746-9
UN:1274
ADR:3,II
:GHS07
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3840 to 1.3860 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
Specification Test Results
Frequently Bought Together