1-Heptadecanol
Catalog # EHERC14122700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Boiling Pt:308.5 °C (1013 hPa)
Melting Pt:53…57 °C
Density:0.967 g/cm³ (20 °C)
:256.47 g/mol
Storage Temperature:Ambient
EINECS:215-932-5
CAS Number:1454-85-9
Formula:H₃C(CH₂)₁₅CH₂OH
MDL Number:MFCD00002822