Order Entry
Austria
Orders LinkContactUsLinkComponent
1-Butanol ≥98.5%, GPR RECTAPUR®

1-Butanol ≥98.5%, GPR RECTAPUR®

Flammable
Irritant
Corrosive
Supplier:   VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
1-Butanol ≥98.5%, GPR RECTAPUR®

4 Options Available Below

Product Details & Documents
Synonyms:  1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
  • CAS Number:71-36-3
  • Formula:H₃C(CH₂)₂CH₂OH
  • MDL Number:MFCD00002964
  • Molecular Weight:74.12 g/mol
  • Merck Index:13,01539
  • Boiling Pt:117.6 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.812 g/cm³ (20 °C)
  • Flash Pt:30 °C
  • Storage Temperature:Ambient
  • EINECS:200-751-6
  • UN:1120
  • ADR:3,III
  • REACH:01-2119484630-38
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 98.5 %
  • Boiling point:
    117 - 119 °C
  • Density (20/4):
    0.807 - 0.812
  • Evaporation residue:
    Max. 100 ppm
Product Family Options

Product Information