1-Butanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.812 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
:M-Clarity™ quality level = MQ300
Flash Pt:30 °C
REACH:01-2119484630-38
:GHS02
:GHS05
:GHS07
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Solubility:77 g/l (20 °C)
- Melting Point:-89 °C
- Molar Mass:74.12 g/mol
- Boiling Point:116 - 118 °C (1013 hPa)
- Vapor Pressure:6.7 hPa (20 °C)
- Flash Point:34 °C
- Refractive Index:1.3993 (20 °C, 589 nm)
- Explosion Limit:1.4 - 11.3 %(V)
- Density:0.81 g/cm3 (20 °C)
- pH:7 (70 g/l, H2O, 20 °C)
- Ignition Point:340 °C
- Evaporation number:33
Specification Test Results
Related Information
Frequently Bought Together