1-Butanol, EMPROVE® ESSENTIAL NF, SAFC®
:71-36-3
:
:H₃C(CH₂)₂CH₂OH
:MFCD00002964
:74.12 g/mol
:13,01539
:117.6 °C (1013 hPa)
:–89 °C
:0.812 g/cm³ (20 °C)
:30 °C
:Ambient
:200-751-6
:1120
:3,III
:01-2119484630-38
:GHS02
:GHS05
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Frequently Bought Together